missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine, 98%, Thermo Scientific Chemicals

Catalog No. p-7054369
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6251506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6251506 Supplier Thermo Scientific Chemicals Supplier No. H6251506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 102185-38-6 | Molecular Formula: C21H34N4O7S | Molecular Weight: 486.584 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102185-38-6
Molecular Formula C21H34N4O7S
Molecular Weight (g/mol) 486.584
MDL Number MFCD00043097
InChI Key CXZHJRGYWGPJSD-HNNXBMFYSA-N
Synonym boc-arg mtr-oh, s-2-tert-butoxycarbonyl amino-5-3-4-methoxy-2,3,6-trimethylphenyl sulfonyl guanidino pentanoic acid, boc-l-arg mtr-oh, ambotzbaa1391, n-boc-n\\'-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, n-alpha-t-butyloxycarbonyl-n'-4-methoxy-2,3,6-trimethylphenyl-sulfonyl-l-arginine, 2s-2-tert-butoxycarbonyl amino-5-n'-4-methoxy-2,3,6-trimethylbenzenesulfonyl carbamimidamido pentanoic acid
PubChem CID 10951013
IUPAC Name (2S)-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)C)OC

Specifications

Color White
Quantity 5 g
Formula Weight 486.53
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only