Learn More
Nalpha-Boc-Nepsilon-Fmoc-L-lysine, 98%
CAS: 84624-27-1 | C26H32N2O6 | 468.55 g/mol
$159.59 - $559.60
Chemical Identifiers
| CAS | 84624-27-1 |
|---|---|
| Molecular Formula | C26H32N2O6 |
| Molecular Weight (g/mol) | 468.55 |
| MDL Number | MFCD00062032 |
| InChI Key | JYEVQYFWINBXJU-ANBDAQEENA-N |
| Synonym | boc-lys fmoc-oh, a-fmoc-l-lysine, n2-boc-n6-fmoc-l-lysine, n-boc-n-fmoc-l-lysine, n-boc-n-fmoc-l-lysine, boclys fmoc, 2s-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, pubchem14939, n, n, a-boc-n |
| PubChem CID | 10983524 |
| SMILES | CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6112606
|
Thermo Scientific Chemicals
H6112606 |
5 g |
Each for $159.59
|
|
|||||
|
AAH6112614
|
Thermo Scientific Chemicals
H6112614 |
25 g |
Each for $559.60
|
|
|||||
Description
Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.
Solubility
Soluble in dimethylformamide. Insoluble in water.
Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 84624-27-1 | |
| 468.55 | |
| JYEVQYFWINBXJU-ANBDAQEENA-N | |
| 10983524 |
| C26H32N2O6 | |
| MFCD00062032 | |
| boc-lys fmoc-oh, a-fmoc-l-lysine, n2-boc-n6-fmoc-l-lysine, n-boc-n-fmoc-l-lysine, n-boc-n-fmoc-l-lysine, boclys fmoc, 2s-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, pubchem14939, n, n, a-boc-n | |
| CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Specifications
| 93°C to 98°C | |
| White | |
| MFCD00062032 | |
| boc-lys fmoc-oh, a-fmoc-l-lysine, n2-boc-n6-fmoc-l-lysine, n2-boc-n6-fmoc-l-lysine, boclys fmoc, n-epsilon-fmoc-alpha-t-boc-l-lysine, n-epsilon-fmoc-alpha-t-boc-l-lysine, pubchem14939, n, n, a-boc-n | |
| JYEVQYFWINBXJU-ANBDAQEENA-N | |
| (2S)-6-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
| 468.55 | |
| Odorless | |
| 98% | |
| Nalpha-Boc-Nepsilon-Fmoc-L-lysine |
| 84624-27-1 | |
| C26H32N2O6 | |
| 3576584 | |
| Soluble in dimethylformamide. Insoluble in water. | |
| CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 5 g | |
| 10983524 | |
| 468.55 | |
| Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only