Learn More
Nalpha-Boc-L-lysine, 97%
CAS: 13734-28-6 | C11H22N2O4 | 246.31 g/mol
Supplier: Thermo Scientific Chemicals H6385922
Description
N-Boc-L-lysine is a protected analogue of a major amino acid participating in the binding of AcH to proteins. N-Boc-L-lysine as well as other t-butoxycarbonyl (Boc) amino acids can be converted to their respective amino acids via cleavage of the urethane bond.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-L-lysine is a protected analogue of a major amino acid participating in the binding of AcH to proteins. N-Boc-L-lysine as well as other t-butoxycarbonyl (Boc) amino acids can be converted to their respective amino acids via cleavage of the urethane bond.
Solubility
Soluble in water. Slightly soluble in methanol.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
| ∼205°C (decomposition) | |
| 13734-28-6 | |
| C11H22N2O4 | |
| 4252546 | |
| Soluble in water. Slightly soluble in methanol. | |
| CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O | |
| 100 g | |
| 2733284 | |
| 97% |
| Nalpha-Boc-L-lysine | |
| White | |
| MFCD00038203 | |
| boc-lys-oh, boc-lysine, n-boc-l-lysine, boc-l-lysine, s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid, nalpha-tert-butoxycarbonyl-l-lysine, n-alpha-tert-butoxycarbonyl-l-lysine, nalpha-boc-l-lysine, boc-lys, n alpha-boc-l-lysine | |
| DQUHYEDEGRNAFO-SVGMAFHSNA-N | |
| (2S)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
| 246.31 | |
| 246.3 | |
| Powder |
Chemical Identifiers
| 13734-28-6 | |
| 246.31 | |
| DQUHYEDEGRNAFO-SVGMAFHSNA-N | |
| 2733284 | |
| CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O |
| C11H22N2O4 | |
| MFCD00038203 | |
| boc-lys-oh, boc-lysine, n-boc-l-lysine, boc-l-lysine, s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid, nalpha-tert-butoxycarbonyl-l-lysine, n-alpha-tert-butoxycarbonyl-l-lysine, nalpha-boc-l-lysine, boc-lys, n alpha-boc-l-lysine | |
| (2S)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
Safety and Handling
EINECSNumber : 237-303-4
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only