missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride, 99+%

C15H22N4O3·HCl

$222.93 - $222.93

Chemical Identifiers

Molecular Formula C15H22N4O3·HCl
MDL Number MFCD00012579
Synonym BAEE
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC105700100
SDS View Documents
Thermo Scientific Chemicals
105700100
10 g Glass Bottle
Each for $222.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

C15H22N4O3·HCl
MFCD00012579
BAEE

Specifications

129°C to 136°C
100.0
99.0% min. (TLC)
99+%
Glass Bottle
MFCD00012579
09, III, 1232
10 g
≥99.0%
Nα-Benzoyl-L-arginine ethyl ester hydrochloride
99.0
White to Yellow
Authentic
C15H22N4O3·HCl
H2NC(=NH)NH(CH2)3CH(NHCOC6H5)CO2C2H5·HCl
−15.0° to −19.0° (24°C, 589nm) (c=2, water)
BAEE
342.83
Powder, Crystals or Granules
Videos
SDS
Documents

Documents

RUO – Research Use Only