missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride, 99+%

Catalog No. AC105700500
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105700500 50 g Glass Bottle
AC105700025 2.5 g Glass Bottle
AC105700100 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC105700500 Supplier Thermo Scientific Chemicals Supplier No. 105700500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C15H22N4O3·HCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula C15H22N4O3·HCl
MDL Number MFCD00012579
Synonym BAEE

Specifications

Melting Point 129°C to 136°C
CAS Min % 99.0
CAS Max % 100.0
Color White to Yellow
Individual Impurity 99.0% min. (TLC)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NHCOC6H5)CO2C2H5·HCl
Beilstein 09, III, 1232
Specific Rotation −15.0° to −19.0° (24°C, 589nm) (c=2, water)
Specific Rotation Condition - 11.50 (24.00°C c=2,H2O)
Quantity 50 g
Formula Weight 342.83
Percent Purity ≥99.0%
Physical Form Powder, Crystals or Granules
Chemical Name or Material Nα-Benzoyl-L-arginine ethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only