missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Acetyl-L-lysine 97.0+%, TCI America™

Catalog No. p-7117775
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21711G 1 g
A21715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21711G Supplier TCI America Supplier No. A21711G

Chemical Identifiers

CAS 1946-82-3
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD00008233
InChI Key VEYYWZRYIYDQJM-JLDDOWRYNA-N
PubChem CID 92907
ChEBI CHEBI:35704
IUPAC Name (2S)-6-amino-2-acetamidohexanoic acid
SMILES CC(=O)N[C@@H](CCCCN)C(O)=O

Specifications

Melting Point 254°C
Color White
Quantity 1 g
Synonym ac-lys-oh, n-alpha-acetyl-l-lysine, n alpha-acetyllysine, nalpha-acetyl-l-lysine, n-acetyl-l-lysine, n2-acetyl-l-lysine, s-2-acetamido-6-aminohexanoic acid, n alpha-acetyl-l-lysine, n 2-acetyl-l-lysine, acetyl-l-lysine
Formula Weight 188.23
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Acetyl-L-lysine