missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Acetyl-D-arginine dihydrate, 98%

CAS: 210545-23-6 | C8H16N4O3 | 216.24 g/mol

$81.66 - $283.78

Chemical Identifiers

CAS 210545-23-6
Molecular Formula C8H16N4O3
Molecular Weight (g/mol) 216.24
MDL Number MFCD00209627
InChI Key SNEIUMQYRCDYCH-ZCFIWIBFSA-N
Synonym ac-d-arg-oh, nalpha-acetyl-d-arginine dihydrate, n-alpha-acetyl-d-arginine dihydrate, 2r-5-carbamimidamido-2-acetamidopentanoic acid dihydrate, n-?-acetyl-d-arginine dihydrate, n-, a-acetyl-d-arginine dihydrate, 2r-5-diaminomethylidene amino-2-acetamidopentanoic acid dihydrate
PubChem CID 72698573
IUPAC Name (2R)-5-{[amino(iminiumyl)methyl]amino}-2-acetamidopentanoate
SMILES CC(=O)N[C@H](CCCNC(N)=[NH2+])C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1856303
View Documents
Thermo Scientific Chemicals
A1856303
1 g
Each for $81.66
Only null left
 
AAA1856306
View Documents
Thermo Scientific Chemicals
A1856306
5 g
Each for $283.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

210545-23-6
216.24
SNEIUMQYRCDYCH-ZCFIWIBFSA-N
72698573
CC(=O)N[C@H](CCCNC(N)=[NH2+])C([O-])=O
C8H16N4O3
MFCD00209627
ac-d-arg-oh, nalpha-acetyl-d-arginine dihydrate, n-alpha-acetyl-d-arginine dihydrate, 2r-5-carbamimidamido-2-acetamidopentanoic acid dihydrate, n-?-acetyl-d-arginine dihydrate, n-, a-acetyl-d-arginine dihydrate, 2r-5-diaminomethylidene amino-2-acetamidopentanoic acid dihydrate
(2R)-5-{[amino(iminiumyl)methyl]amino}-2-acetamidopentanoate

Specifications

∼270°C
C8H16N4O3
ac-d-arg-oh, nalpha-acetyl-d-arginine dihydrate, n-alpha-acetyl-d-arginine dihydrate, 2r-5-carbamimidamido-2-acetamidopentanoic acid dihydrate, n-?-acetyl-d-arginine dihydrate, n-, a-acetyl-d-arginine dihydrate, 2r-5-diaminomethylidene amino-2-acetamidopentanoic acid dihydrate
CC(=O)N[C@H](CCCNC(N)=[NH2+])C([O-])=O
1 g
72698573
99%
210545-23-6
MFCD00209627
SNEIUMQYRCDYCH-ZCFIWIBFSA-N
(2R)-5-{[amino(iminiumyl)methyl]amino}-2-acetamidopentanoate
216.24
252.29 (216.25 Anhydrous)
Nalpha-Acetyl-D-arginine dihydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-225-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only