missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Acetyl-D-arginine dihydrate, 98%

Catalog No. AAA1856303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1856303 1 g
AAA1856306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1856303 Supplier Thermo Scientific Chemicals Supplier No. A1856303

CAS: 210545-23-6 | C8H16N4O3 | 216.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 210545-23-6
Molecular Formula C8H16N4O3
Molecular Weight (g/mol) 216.24
MDL Number MFCD00209627
InChI Key SNEIUMQYRCDYCH-ZCFIWIBFSA-N
Synonym ac-d-arg-oh, nalpha-acetyl-d-arginine dihydrate, n-alpha-acetyl-d-arginine dihydrate, 2r-5-carbamimidamido-2-acetamidopentanoic acid dihydrate, n-?-acetyl-d-arginine dihydrate, n-, a-acetyl-d-arginine dihydrate, 2r-5-diaminomethylidene amino-2-acetamidopentanoic acid dihydrate
PubChem CID 72698573
IUPAC Name (2R)-5-{[amino(iminiumyl)methyl]amino}-2-acetamidopentanoate
SMILES CC(=O)N[C@H](CCCNC(N)=[NH2+])C([O-])=O

Specifications

Melting Point ∼270°C
Quantity 1 g
Formula Weight 252.29 (216.25 Anhydrous)
Percent Purity 99%
Chemical Name or Material Nalpha-Acetyl-D-arginine dihydrate
EINECSNumber 219-225-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only