missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine 98.0+%, TCI America™
$29.83 - $218.10
Chemical Identifiers
| CAS | 71989-38-3 |
|---|---|
| Molecular Formula | C28H29NO5 |
| Molecular Weight (g/mol) | 459.54 |
| MDL Number | MFCD00037129 |
| InChI Key | JAUKCFULLJFBFN-KSYWNVGFNA-N |
| Synonym | fmoc-tyr tbu-oh, fmoc-o-tert-butyl-l-tyrosine, fmoc-tyr but-oh, fmoc-tyr t-bu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, fmoc-tyr but, n-fmoc-o-tert-butyl-l-tyrosine, nalpha-fmoc-o-tert-butyl-l-tyrosine, n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine, 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid |
| PubChem CID | 10895791 |
| IUPAC Name | (2S)-3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid |
| SMILES | CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1 |
Chemical Identifiers
| 71989-38-3 | |
| 459.54 | |
| JAUKCFULLJFBFN-KSYWNVGFNA-N | |
| 10895791 | |
| CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1 |
| C28H29NO5 | |
| MFCD00037129 | |
| fmoc-tyr tbu-oh, fmoc-o-tert-butyl-l-tyrosine, fmoc-tyr but-oh, fmoc-tyr t-bu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, fmoc-tyr but, n-fmoc-o-tert-butyl-l-tyrosine, nalpha-fmoc-o-tert-butyl-l-tyrosine, n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine, 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid | |
| (2S)-3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid |
Specifications
| 71989-38-3 | |
| White | |
| MFCD00037129 | |
| fmoc-tyr tbu-oh, fmoc-o-tert-butyl-l-tyrosine, fmoc-tyr but-oh, fmoc-tyr t-bu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, fmoc-tyr but, n-fmoc-o-tert-butyl-l-tyrosine, nalpha-fmoc-o-tert-butyl-l-tyrosine, n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine, 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid | |
| CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1 | |
| 459.54 | |
| 459.54 | |
| Crystalline Powder |
| 153°C | |
| C28H29NO5 | |
| 1 g | |
| JAUKCFULLJFBFN-KSYWNVGFNA-N | |
| (2S)-3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid | |
| 10895791 | |
| ≥98.0% (T,HPLC) | |
| Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator