missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 153°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 459.54 |
| Percent Purity | ≥98.0% (T,HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |