missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-asparagine 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 354.36 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-asparagine |
Safety and Handling
| TSCA | No |