missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-(5-Fluoro-2,4-dinitrophenyl)-L-leucinamide 98.0+%, TCI America™
Supplier: TCI America A5523100MG
| Quantity | 100 mg |
|---|
Chemical Identifiers
| 178065-29-7 | |
| 314.273 | |
| WCOZOJGXDVGGIK-VIFPVBQESA-N | |
| (2S)-2-(5-fluoro-2,4-dinitroanilino)-4-methylpentanamide |
| C12H15FN4O5 | |
| MFCD03844761 | |
| 15817226 | |
| CC(C)CC(C(=O)N)NC1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F |
Specifications
| 170°C | |
| 178065-29-7 | |
| C12H15FN4O5 | |
| 1325 | |
| CC(C)CC(C(=O)N)NC1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F | |
| 100 mg | |
| 15817226 | |
| ≥98.0% (HPLC) |
| Nalpha-(5-Fluoro-2,4-dinitrophenyl)-L-leucinamide [HPLC Labeling Reagent for e.e. Determination] | |
| Yellow | |
| MFCD03844761 | |
| WCOZOJGXDVGGIK-VIFPVBQESA-N | |
| (2S)-2-(5-fluoro-2,4-dinitroanilino)-4-methylpentanamide | |
| 314.273 | |
| 314.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No