missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-(5-Fluoro-2,4-dinitrophenyl)-D-leucinamide 98.0+%, TCI America™
$254.41 - $1,346.11
Chemical Identifiers
| CAS | 178065-30-0 |
|---|---|
| Molecular Formula | C12H15FN4O5 |
| Molecular Weight (g/mol) | 314.27 |
| MDL Number | MFCD03844762 |
| InChI Key | WCOZOJGXDVGGIK-UHFFFAOYNA-N |
| Synonym | Nalpha-(2,4-Dinitro-5-fluorophenyl)-D-leucinamide |
| PubChem CID | 15817227 |
| IUPAC Name | 2-[(5-fluoro-2,4-dinitrophenyl)amino]-4-methylpentanamide |
| SMILES | CC(C)CC(NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(N)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
A5524100MG
|
N/A | 100 mg |
Each for $254.41
|
|
|||||
|
A55241G
|
N/A | 1 g |
Each for $1,346.11
|
|
|||||
Chemical Identifiers
| 178065-30-0 | |
| 314.27 | |
| WCOZOJGXDVGGIK-UHFFFAOYNA-N | |
| 15817227 | |
| CC(C)CC(NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(N)=O |
| C12H15FN4O5 | |
| MFCD03844762 | |
| Nalpha-(2,4-Dinitro-5-fluorophenyl)-D-leucinamide | |
| 2-[(5-fluoro-2,4-dinitrophenyl)amino]-4-methylpentanamide |
Specifications
| 170°C | |
| Yellow | |
| MFCD03844762 | |
| Nalpha-(2,4-Dinitro-5-fluorophenyl)-D-leucinamide | |
| CC(C)CC(NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(N)=O | |
| 314.27 | |
| 314.27 | |
| Crystalline Powder |
| 178065-30-0 | |
| C12H15FN4O5 | |
| 1325 | |
| WCOZOJGXDVGGIK-UHFFFAOYNA-N | |
| 2-[(5-fluoro-2,4-dinitrophenyl)amino]-4-methylpentanamide | |
| 15817227 | |
| ≥98.0% (HPLC) | |
| Nalpha-(5-Fluoro-2,4-dinitrophenyl)-D-leucinamide [HPLC Labeling Reagent for e.e. Determination] |
Safety and Handling
TSCA : No