missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
$464.27 - $464.27
Chemical Identifiers
| CAS | 156648-40-7 |
|---|---|
| Molecular Formula | C31H36N2O6 |
| Molecular Weight (g/mol) | 532.64 |
| MDL Number | MFCD00467662 |
| InChI Key | CNDRLQTWKWIHEP-KSYWNVGFNA-N |
| Synonym | dde-lys fmoc-oh, a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-, dde-lysine fmoc-oh, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino hexanoic acid, dde-lysine fmoc-oh, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino hexanoic acid, 2s-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n-alpha-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-epsilon-fmoc-l-lysine, n-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n'-fmoc-l-lysine, ambotzdaa1015, n-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n'-fmoc-l-lysine, n- |
| PubChem CID | 51340473 |
| IUPAC Name | (2S)-2-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethylamino]-6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid |
| SMILES | CC(N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6644603
|
Thermo Scientific Chemicals
H6644603 |
1 g |
Each for $464.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 156648-40-7 | |
| 532.64 | |
| CNDRLQTWKWIHEP-KSYWNVGFNA-N | |
| 51340473 | |
| CC(N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O |
| C31H36N2O6 | |
| MFCD00467662 | |
| dde-lys fmoc-oh, a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-, dde-lysine fmoc-oh, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino hexanoic acid, dde-lysine fmoc-oh, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino hexanoic acid, 2s-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n-alpha-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-epsilon-fmoc-l-lysine, n-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n'-fmoc-l-lysine, ambotzdaa1015, n-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n'-fmoc-l-lysine, n- | |
| (2S)-2-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethylamino]-6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid |
Specifications
| 156648-40-7 | |
| C31H36N2O6 | |
| dde-lys fmoc-oh, a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-, dde-lysine fmoc-oh, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino hexanoic acid, dde-lysine fmoc-oh, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino hexanoic acid, 2s-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n-alpha-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-epsilon-fmoc-l-lysine, n-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n'-fmoc-l-lysine, ambotzdaa1015, n-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n'-fmoc-l-lysine, n- | |
| CC(N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O | |
| 1 g | |
| 51340473 | |
| 98% | |
| Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine |
| White | |
| MFCD00467662 | |
| CNDRLQTWKWIHEP-KSYWNVGFNA-N | |
| (2S)-2-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethylamino]-6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid | |
| 532.64 | |
| 532.64 | |
| Powder |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only