missing translation for 'onlineSavingsMsg'
Learn More

Naloxone Hydrocloride, MP Biomedicals™

Catalog No. p-7139870
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219024583 250 mg
MP219024591 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219024583 Supplier MP Biomedicals Inc Supplier No. 0219024583
Only null left

Naloxone Hydrocloride Off-white Crystalline Powder

  • The role of naloxone (1μM) and other opioids on apoptosis in several human cancer cell lines has been probed. Naloxone (10-100μM) has been used to study proliferation and neurogenesis in cultured rat adult hippocampal progenitors.
  • Naloxone has also been utilized to modulate the NMDA receptor activation capability of theμ-opioid receptor agonist DAMGO. It displaces morphine and other opiate agonists from the opiate receptor.
  • It is more potent than Nalorphine and is also an effective antagonist for mixed agonist-antagonist drugs such as pentazorcine. Naloxone has no respiratory depressant activity.

Chemical Identifiers

CAS 357-08-4
Molecular Formula C19H22ClNO4
Molecular Weight (g/mol) 363.84
InChI Key RGPDIGOSVORSAK-STHHAXOLSA-N
Synonym Narcan, EN-1530, (5α)-4,5-Epoxy-3,14-dihydroxy-17-(2-propenyl)morphinan-6-one hydrochloride, 17-allyl-4,5α- epoxy-3,14-dihydroxymorphinan-6-one hydrochloride

Specifications

Melting Point 129.0°C to 129.2°C (literature)
Color Off-white
Quantity 250 mg
Beilstein 6260807
Solubility Information Soluble in water (50mg/mL - clear, colorless to faint yellow solution), ethanol (3.3mg/mL), or methanol (50mg/mL - clear, colorless solution).
Physical Form Crystalline Powder
RTECSNumber QD2275000
storageNote1 Store at +4°C, Store Desiccated.