missing translation for 'onlineSavingsMsg'
Learn More

Nalidixic acid sodium salt

Catalog No. AC463550010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463550010 1 g
AC463550050 5 g
AC463550250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC463550010 Supplier Thermo Scientific Chemicals Supplier No. 463550010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3374-05-8 | C12H11N2NaO3 | 254.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5-8-3374
Molecular Formula C12H11N2NaO3
Molecular Weight (g/mol) 254.22
InChI Key ROKRAUFZFDQWLE-UHFFFAOYSA-M
IUPAC Name sodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate
SMILES [Na+].CCN1C=C(C([O-])=O)C(=O)C2=CC=C(C)N=C12

Specifications

Melting Point >280.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: 50 g/L (20°C).
Formula Weight 254.22
Chemical Name or Material Nalidixic acid sodium salt
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

May cause an allergic skin reaction.

May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation or rash occurs: Get medical advice/attention.

In case of inadequate ventilation wear respiratory protection.

IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.

EINECSNumber 222-159-7

RUO – Research Use Only