missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles

Keep hazardous volatile liquids off the lab bench. Unitary wash bottles feature a unique vent system, and eliminate draw tube assembly and associated risk of contamination.

Supplier:  Thermo Scientific™ 24360504

 View more versions of this product

Catalog No. 03-409-20D



Only null left

Description

Description

Keep hazardous volatile liquids off the lab bench with Thermo Scientific™ Nalgene™ Vented Unitary Right-To-Know LDPE Wash Bottles, featuring a unique vent system. Unitary bottle prevents contamination when filling the bottle; no draw tube assembly to pick up contamination off the lab bench. A strong, steady, no sputter stream of liquid is dispensed when the wash bottle is squeezed. Marked with four color, NFPA diamonds, GHS compliant hazard symbols, and right-to-know chemical information. Choose from seven chemical markings.

  • Bottle and tubulation molded in one piece to prevent chemical splash when removing closure and contamination when filling
  • Unique 38mm colored vented closure designed to prevent dripping
  • Sturdy polypropylene closures (HDPE on Sodium Hypochloride bottles)
  • Color-coded bar corresponds to closure, preventing cross-contamination
  • Prelabeled with ICS name and formula, as well as NFPA symbols for chemical handling, hazard, care and storage of six commonly-used chemicals
  • Leakproof†
Specifications

Specifications

16 oz., 500 mL
16 oz.
RTK Wash Bottle
24
Yellow
Meet OSHA requirements for workplace chemical container labeling.
No
Cylindrical
Isopropanol
Screw Cap
Not Autoclavable
500 mL
Translucent
LDPE
4
Yes
Yes
Narrow
Wash Bottle
Polypropylene
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions