Learn More
Thermo Scientific™ Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Keep hazardous volatile liquids off the lab bench. Unitary wash bottles feature a unique vent system, and eliminate draw tube assembly and associated risk of contamination.
Supplier: Thermo Scientific™ 24360502
Description
Keep hazardous volatile liquids off the lab bench with Thermo Scientific™ Nalgene™ Vented Unitary Right-To-Know LDPE Wash Bottles, featuring a unique vent system. Unitary bottle prevents contamination when filling the bottle; no draw tube assembly to pick up contamination off the lab bench. A strong, steady, no sputter stream of liquid is dispensed when the wash bottle is squeezed. Marked with four color, NFPA diamonds, GHS compliant hazard symbols, and right-to-know chemical information. Choose from seven chemical markings.
- Bottle and tubulation molded in one piece to prevent chemical splash when removing closure and contamination when filling
- Unique 38mm colored vented closure designed to prevent dripping
- Sturdy polypropylene closures (HDPE on Sodium Hypochloride bottles)
- Color-coded bar corresponds to closure, preventing cross-contamination
- Prelabeled with ICS name and formula, as well as NFPA symbols for chemical handling, hazard, care and storage of six commonly-used chemicals
- Leakproof†
Specifications
| 16 oz., 500 mL | |
| 16 oz. | |
| RTK Wash Bottle | |
| 24 | |
| White | |
| Meet OSHA requirements for workplace chemical container labeling. | |
| No | |
| Cylindrical | |
| Ethyl Alcohol | |
| Screw Cap | |
| Not Autoclavable |
| 500 mL | |
| Translucent | |
| LDPE | |
| 4 | |
| Yes | |
| Yes | |
| Narrow | |
| Wash Bottle | |
| Polypropylene | |
| No |