missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks

Catalog No. 02923151
Change view
Click to view available options
For Use With (Equipment):
125, 250 mL flasks
500, 1000, 2000 mL flasks
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
02923151 500, 1000, 2000 mL flasks
02923150 125, 250 mL flasks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02923151 Supplier Thermo Scientific™ Supplier No. 41140045

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ensure a sterile gas exchange for shaker and suspension cell culture by using Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks.

Ensure a sterile gas exchange for shaker and suspension cell culture by using Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks.

  • Blue high-density polyethylene with hydrophobic 0.2 μm PTFE membrane
  • Air exchange without unscrewing closure
  • Membrane heat welded into place for effective sealing

Warranty: 90 days

Specifications

Color Blue
Material HDPE
For Use With (Equipment) 500, 1000, 2000 mL flasks
No. per Case 12/Cs.
No. per Pack 1 Ea.
Product Type Flask Closure
Autoclavable Not Autoclavable
Disposable Single-use
Sterility Sterile
Closure Type Vented

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.