Learn More
Thermo Scientific™ Nalgene™ Teflon™ Low Particulate Bottles
Nalgene Teflon Low Particulate Bottles are certified clean to USP <788> standards, making them an ideal solution for biological and pharmaceutical manufacturing.
$3230.07 - $6344.21
Specifications
| Color | Clear |
|---|---|
| For Use With (Application) | Storage, Transportation â |
| Material | Teflon FEP |
| No. per Pack | 1 Ea. |
| Sterility | Non-sterile |
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
|
03-313-104
|
Thermo Scientific™
3816000004 |
125 mL | 24 mm |
Case of 6 Each for $3,230.07
|
|
||||
|
03-313-105
|
Thermo Scientific™
3816000008 |
250 mL | 24 mm |
Case of 4 Each for $3,649.96
|
|
||||
|
03-313-106
|
Thermo Scientific™
3816000016 |
500 mL | 28 mm |
Case of 4 Each for $4,766.18
|
|
||||
|
03-313-107
|
Thermo Scientific™
3816000032 |
1 L | 38 mm |
Case of 4 Each for $6,344.21
|
|
||||
Description
Sample and store active pharmaceutical ingredients and bulk intermediates optimally with Thermo Scientific™ Nalgene™ Low Particulate/Low Metals Bottles Made of Teflon™ FEP with Closure, certified to contain less than 20 particles per mL at 0.3 μm and greater. Metals certified to micrograms per liter (ppb) levels. Excellent for storing high purity chemicals, this product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.
Features:
- Certified to particulate and trace metals levels
- Linerless Tefzel™ ETFE closures provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks, or contaminate your reagents
- Exhibits flexibility, impact resistance and excellent visibility of contents for a wide variety of uses in the lab
- Excellent chemical resistance of almost any lab chemical
- Excellent for storing high purity chemicals
- Non-sterile/autoclavable
- Translucent
Each bottle is double bagged under Class 10 laminar flow hoods inside a Class 100 clean room. Excellent for ICP-MS reagent and standard storage. Product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.
Metals certified to microliter (ppb) levels: <0.20 Hg, <0.5 Be, <1.0 As/Cd/Pb, <2.0 Sb/Se, <5.0 Ag/Co/Cr/Cu/Mn/Tl/V, <10 Ba/Ni/Zn, <50 Mg, <75 Al, <100 Ca/Fe/K/Na
Warranty: 90 days
Specifications
| Clear | |
| Teflon FEP | |
| Non-sterile | |
| White | |
| Certified to particulate and trace metals levels. Metals certified to microliter (ppb) levels <0.20 Hg, <0.5 Be, <1.0 As/Cd/Pb, <2.0 Sb/Se, <5.0 Ag/Co/Cr/Cu/Mn/Tl/V, <10 Ba/Ni/Zn, <75 Al, <100 Ca/Fe/K/Na. Certified to contain <20 particles | |
| Narrow | |
| Low Particulate/Low Metals Bottle | |
| Tefzel™ ETFE | |
| No | |
| Upstream process solution storage. Upstream Validation. Downstream process solution storage. Downstream Validation |
| Storage, Transportation â | |
| 1 Ea. | |
| -100°C to 260°C | |
| USP <788> (Low particulate) | |
| Yes | |
| Round | |
| Unlined | |
| Screw Cap | |
| Autoclavable |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.