missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Self-Zeroing Buret Filler Kit
GREENER_CHOICE

Catalog No. 03860
Change view
Click to view available options
For Use With (Equipment):
25 or 50 mL Burette
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03860 25 or 50 mL Burette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03860 Supplier Thermo Scientific™ Supplier No. 36452550
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Save time and increase convenience. This filler kit converts any conventional glass or plastic 25mL or 50mL buret into an automatic-filling, self-zeroing buret.

Save time and increase convenience. The Thermo Scientific™ Nalgene™ Self-Zeroing Buret Filler Kit converts any conventional glass or plastic 25mL or 50mL buret into an automatic-filling, self-zeroing buret. Avoid messy spills from manual fi

Operating Instructions:

The polypropylene fill tube connects to the end of the transfer tubing and is inserted into the mouth of the buret using the provided adapter. Adjust the fill tube tip so it is just above the zero mark of your buret. Squeeze the bottle to fill the buret above the zero mark. When you release the squeeze on the bottle, the excess titrant will be drawn back into the reservoir bottle leaving the titrant level in the buret at the zero line.

Includes:

  • 1000mL low-density polyethylene squeeze bottle
  • 4′ length (1.2m) of clear, flexible PVC tubing
  • Fill tube
  • Adapter
  • Closure for reagent bottle.
Buret not included.

Specifications

For Use With (Equipment) 25 or 50 mL Burette
Material LDPE
No. per Pack 1
No. per Case 4
Includes 1000 mL Squeeze Bottle, 4 ft. PVC Tubing, Fill Tube, Adapter and Closure
Type Buret Filler Kit

Buret not included