Learn More
Thermo Scientific™ Nalgene™ Sanitary 1 in. Hose Barb Connector

Nalgene Sanitary Tri-clamp Hose barb fitting supports connections between 1 in. sanitary fitting to flexible tubing with inner diameter of 0.5 in..
Supplier: Thermo Scientific™ 72110500
Description
Thermo Scientific™ Nalgene Sanitary 1 in. Hose Barb connector supports connections between 1 in. sanitary fitting to flexible tubing with inner diameter of 0.5 in..
- Polypropylene
- Environmental benefits include:
- Renewable Energy
Specifications
0.5 in. | |
Polypropylene | |
0.5 in. (13mm) |
13 mm | |
2/Pk. | |
Hose Barb Connector |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.