Learn More
Thermo Scientific™ Nalgene™ Repairable Hand-Operated PVC Vacuum Pumps with Gauge
GREENER_CHOICE
Catalog No. 13874612A
Change view
Click to view available options
Flow Rate:
15 cc/stroke
36 cc/stroke
For Use With:
6132-2000 Repair Kit
6132-3000 Repair Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate For Use With
13874612A 15 cc/stroke 6132-2000 Repair Kit
13874612B 36 cc/stroke 6132-3000 Repair Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13874612A Supplier Thermo Scientific™ Supplier No. 61320010
Easily operate these repairable hand-operated PVC vacuum pumps, ideal for siphoning, filtration, liquid transfer, bleeding fluid lines and checking for leaks.
Easily operate Thermo Scientific™ Nalgene™ Repairable Hand-Operated PVC Vacuum Pumps with Gauge, ideal for siphoning, filtration, liquid transfer, bleeding fluid lines and checking for leaks. Lightweight units are sealed and self-lubricating with removable covers on exhaust port and trigger to release vacuum without disconnecting pumps from the line. Quickly attain and hold vacuum of 25 in. (635 mm) Hg. The vacuum trigger release requires only one hand to operate and releases vacuum with the touch of an index finger.
Features:
  • Gauge monitors vacuum in kPa and inches of mercury for visual indication of pump rate and progress.
  • New handle design is more durable and ergonomic.
  • Adjustable vacuum release rate provides control.
  • Full blank port pressure is obtained with only two strokes, making operation efficient.
  • Removable cover on exhaust port and trigger allow release vacuum without disconnecting pumps from the line.
  • Adjustable vacuum release rate is controlled by a trigger on the pump.
  • Nozzle fits standard 0.25″ I.D. tubing.
  • System can be disassembled for repair and replacement of internal parts.
Includes:
2′ (0.6 m) Nalgene 180 clear plastic tubing
Specifications
Description Nalgene repairable PVC hand pump with gauge
No. per Pack 1
No. per Case 4
Max. Vacuum 25 in. Hg
Flow Rate 15 cc/stroke
Type Vacuum Pump
For Use With 6132-2000 Repair Kit