missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
GREENER_CHOICE

Catalog No. 0296310A
Change view
Click to view available options
Capacity (Metric):
9 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
0296310A 9 L
0296310C 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0296310A Supplier Thermo Scientific™ Supplier No. 23220020
Get a Water Bottle with Plastics Purchase
View all promotions

For Use With: Ideal for use when visual inspection of contents is necessary; Autoclavable; Closure Material: Polypropylene; Color: Clear

Easily fill and clean the Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ with Spigot, which features a wide-mouth opening, a sturdy stainless-steel handle and space-saving rectangular design. Clear construction allows easy viewing of carboy contents. Super tough; impact resistant. Ideal for refrigerated or frozen storage of aqueous solutions, or for dispensing sterile water, aqueous reagents and media. Nalgene polycarbonate carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features

  • Molded-in graduations in 1 gallon and 5L increments for ease in dispensing
  • Stainless-steel handle for ease in transporting containers
  • Molded-in spigot boss provides strength and leak resistance
  • A clear wall makes this carboy ideal when visual inspection of contents is necessary
  • Follow autoclave instructions provided with the product for proper function; tubing adapter is not autoclavable
  • Autoclavable/Transparent/Graduated

Includes:

Polypropylene spigot and screw closure

Specifications

Autoclavable Autoclavable
Capacity (English) 2.4 gal.
Capacity (Metric) 9 L
Closure Material Polypropylene
Closure Size 100 mm
Color Transparent
Description Autoclavable Clear Rectangular Carboy w/Spigot
Diameter (Metric) Inner Neck 86 mm
For Use With (Application) Ideal for use when visual inspection of contents is necessary
Height (Metric) With Closure 361 mm
Includes Molded-in spigot boss w/tubing adapter
Length (Metric) 220 mm
Material Polycarbonate
No. per Case 6/Cs.
No. per Pack 1 Ea.
Product Type Carboy with Spigot
Width (Metric) 153 mm
Show More Show Less