Learn More
Thermo Scientific™ Nalgene™ PVDF True Union Clamps, 0.75 inch
GREENER_CHOICE
Use Thermo Scientific™ Nalgene™ PVDF True Union Clamps with silicone gasket and end caps to experience reliable, repeatable seals.
Supplier:  Thermo Scientific™ 26700075
Catalog No. 14831318A
Only null left
Description
Description
Use Thermo Scientific™ Nalgene™ PVDF True Union Clamps with silicone gasket and end caps to experience reliable, repeatable seals. Molded of PVDF fluoride, these threaded clamps are a fully autoclavable clamp system.
  • 0.75 in. Mini Clamp is threaded and autoclavable
  • Recommended for use with Nalgene™ Sanitary Carboys (Cat. Nos. 2261, 2630 and 2640) and Nalgene™ End Caps (Cat. Nos. 2688 and 2689)
  • Recommended for use with Nalgene™ Autoclavable Carboys with Spigot (Cat. No. 2319)
Specifications
Specifications
1/Pk., 1/Cs.
Nalgene™ Sanitary Carboys, Nalgene™ Autoclavable Carboys with Spigot
0.75 in.
PVDF
Mini
Product Suggestions
Product Suggestions