missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes

Description
Simplify measurements using Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes, which feature molded-in graduations. Translucent polypropylene copolymer (PPCO) has excellent chemical resistance.
Features:
- Translucent Polypropylene Copolymer (PPCO) is compatible with a wide range of lab reagents
- Designed for low-speed centrifugation up to 6000 × g in refrigerated or non-refrigerated centrifuges (tubes must be filled at least 80% for proper performance)
- Conical bottoms concentrate pellet in a small area for easy isolation and retrieval
- Molded-in graduations last the life of the tube
- Last longer than polycarbonate tubes under conditions of repeated autoclaving
- Tube are both autoclavable and graduated
Ordering Information:
Supplied without closure; friction fit closures available. For 15mL use DS3111-00017; for 50mL use DS3111-0029.
Specifications
Specifications
| Capacity (Metric) | 15 mL |
| Material | Polypropylene Copolymer |
| No. per Case | 100 |
| No. per Pack | 10 |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Shape | Conical |
| Length (Metric) | 118.6 mm |
| Bottom Shape | Conical |
| Writing Area | No |
| Show More |