missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Copolymer (PPCO) Separatory Funnels with Closure
GREENER_CHOICE

Catalog No. 1043730D
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
1043730D 1000 mL
1043730A 125 mL
1043730B 250 mL
1043730C 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1043730D Supplier Thermo Scientific™ Supplier No. 43001000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform a variety of lab duties with chemically inert, translucent and autoclavable polypropylene separatory funnels.

Perform a variety of liquid phase separations with translucent and autoclavable Thermo Scientific™ Nalgene™ Polypropylene Copolymer (PPCO) Separatory Funnels. Good chemical compatibility with a wide range of laboratory solutions involved in liquid-liquid extractions. Polypropylene closure and TFE stopcock require no lubrication. These separatory funnels are impact resistant and won't chip or shatter, making them a valuable asset to lab safety programs.

Features:

  • Separation lines between immiscible liquids are easily observed
  • Phase interface is observable down to the stopcock
  • TFE stopcocks need no lubrication, allowing for easier handling and operation
  • Funnel is chemically compatible with a wide range of lab reagents
  • Autoclavable/Leakproof

Includes:

  • Polypropylene funnel, Teflon™ TFE stopcock and housing, polypropylene screw closure
Ordering Information:
  • For replacement stopcock, order Cat. No. 714301-1025
Notes:
  • Funnels are susceptible to softening and swelling from extended exposure to aggressive solvents though the effect is reversible.
  • Stem lengths, 65mm. Autoclavable except for stopcock assembly, which is easily removed for chemical sterilization.
  • Autoclavable except for stopcock assembly, which is easily removed and chemically sterilized.

Specifications

Capacity (Metric) 1000 mL
Material Polypropylene Copolymer (PPCO)
Height (Metric) 371 mm
Description 53mm closure
Quantity 1/Pk.; 4/Cs.
Product Type Separatory Funnel

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.