missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene, Rectangular Carboy with Closure
GREENER_CHOICE

Catalog No. 0296153C
Change view
Click to view available options
Capacity (Metric):
9 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
0296153C 20 L
0296153A 9 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0296153C Supplier Thermo Scientific™ Supplier No. 22120050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use With: Store solutions and handle large volumes of powders and other solid samples; Autoclavable; Closure Material: Polypropylene; Color: White

Store solutions and handle large volumes of powders and other solid samples with Thermo Scientific™ Nalgene™ Rectangular Polypropylene Carboys with Closure. Featuring a space-saving design, a wide mouth opening for filling and cleaning and a sturdy stainless-steel handle, these carboys are tough, translucent and autoclavable.

Features:

  • Wide-mouth opening for easy filling, transferring and cleaning of carboys
  • Molded in graduations in 1 gallon and 5 L increments for ease in dispensing
  • Stainless-steel handle for ease in transporting containers
  • Autoclavable, graduated and leakproof†

Includes:

  • Polypropylene screw closure

Specifications

Autoclavable Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Closure Size 100-415 mm
Color White
Dimensions (L x W x H) 13 x 9 x 16 in. (320 x 229 x 399 mm)
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Material Polypropylene
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Rectangular Carboy
Sterility Non-sterile
Workflow Step Upstream process solution storage. Upstream Validation
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.