Learn More
Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp
Description
From completely open to restricted flow to completely closed—experience precise, one-hand regulation of fluid flow with the Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp. This plastic clamp will not rust, corrode or deform. Molded in one piece with no sharp edges, it accommodates a wide range of tubing.
Specifications
Specifications
| Quantity | 72/Cs. |
| Type | Pinch Clamp |
| Material | Polypropylene |
| For Use With (Equipment) | Fits tubing with O.D.: 1/4 to 7/16 in. |
| Description | Pinch clamp; Fits tubing with O.D.: 1/4 to 7/16 in. |