missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene, Heavy-Duty Vacuum Carboy with Handle
GREENER_CHOICE

Catalog No. 0296015
Change view
Click to view available options
Capacity (Metric):
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
0296015 20 L
0296014 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0296015 Supplier Thermo Scientific™ Supplier No. 22260050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use With: Ideal for the most extreme service conditions; Autoclavable; Closure Material: Polypropylene; Color: White

Ideal for the most extreme service conditions, the Thermo Scientific™ Nalgene™ Polypropylene Heavy Duty Vacuum Carboy is useful as a vacuum trap and will hold a full vacuum up to eight hours. Constructed of thick-walled natural polypropylene to provide added strength, it is supplied with a white 83B PP closure with thermoplastic elastomer (TPE) gasket. Customization options available for fluid transfer applications. Fit perfectly with a variety of optional 83B closures with standard tubing sets (such as Nalgene™ Top Works™, Nalgene™ Filling/Venting) and custom tubing set designs.

  • Autoclavable when closure threads are disengaged and vessel is empty
  • Not rated for pressure use

Ordering Information:

  • For closure options in vacuum applications, use with Nalgene Quick Filling/Venting Closures (Cat. No. 2158), or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulkhead Fittings (Cat. No. 6149)
  • For 1 to 5 L sizes, please refer to Nalgene Heavy Duty Vacuum Bottles (Cat. No. 2126)

Specifications

Autoclavable Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Closure Size 83B
Color White
Diameter (Metric) Outer 282 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Includes Heavy-Duty Vacuum Carboy with Handles
Material Polypropylene
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Carboy with Handle
Sterility Non-sterile
Volume (English) Brim 5.5 gal.
Volume (Metric) Brim 22.5 L
Workflow Step Upstream process solution storage. Upstream Validation
Show More Show Less