missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Combine these flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
Supplier: Thermo Scientific™ 41021000
Description
Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
- Translucent wall makes liquid level easily visible
- 2000mL size has molded-in graduations
- Mouth sized to accept standard sized stoppers (stopper sold separately)
- Repeatedly autoclavable for long life and rigorous use
Specifications
| 32 oz. | |
| 1000 mL | |
| 6 | |
| Autoclavable | |
| No | |
| Non-sterile | |
| Stopper No. 8 |
| Flask Erlenmeyer PP 1000mL | |
| PPCO | |
| 2 | |
| Reusable | |
| Narrow | |
| Plain |