missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
GREENER_CHOICE

Catalog No. 1004110F
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
1000 mL
2000 mL
Closure Size:
Stopper No. 10
Stopper No. 3
Stopper No. 6
Stopper No. 8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
1004110F 2000 mL Stopper No. 10
1004110B 125 mL Stopper No. 3
1004110C 250 mL Stopper No. 6
1004110E 1000 mL Stopper No. 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1004110F Supplier Thermo Scientific™ Supplier No. 41022000
Get a Water Bottle with Plastics Purchase
View all promotions

Combine these flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.

Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.

  • Translucent wall makes liquid level easily visible
  • 2000mL size has molded-in graduations
  • Mouth sized to accept standard sized stoppers (stopper sold separately)
  • Repeatedly autoclavable for long life and rigorous use

Specifications

Capacity (English) 64 oz.
Description Flask Erlenmeyer PP 2000mL
Capacity (Metric) 2000 mL
Material PPCO
No. per Case 6
No. per Pack 2
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Plain
Closure Size Stopper No. 10
Show More Show Less