missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene, Carboy with Tubulation
GREENER_CHOICE

Catalog No. 0269023
Change view
Click to view available options
No. per Pack:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
0269023 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Completely disengage threads or remove closure before autoclaving. Not recommended for hazardous materials.
Catalog No. 0269023 Supplier Thermo Scientific™ Supplier No. 23010050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Completely disengage threads or remove closure before autoclaving. Not recommended for hazardous materials.

For Use With: For use with distilled water and culture media.; Autoclavable; Closure Material: Polypropylene; Color: White

Ensure more complete drainage with the low outlet of Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with Bottom Tubulation, excellent for distilled water and culture media.

Features:
  • Bottom tubulation molded in one piece with bottle
  • Molded-in handles for ease in transporting containers
  • Molded-in graduations in 1 gal. and 5L increments
  • Neck accepts No. 13.5 rubber stopper
  • Autoclavable/Graduated/Leakproof† when used with the provided Nalgene closures

Ordering Information:

Both sizes accept 0.5in. I.D. (1.3cm) tubing - Not Included

Includes:

White polypropylene screw cap with TPE gasket

Note:

A hose clamp should be used when attaching tubing to ensure tight fit.

Specifications

Autoclavable Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Closure Size 83B
Color White
Description Autoclavable Carboy w/Bottom Tubulation
Diameter (Metric) Inner Neck 64 mm
Diameter (Metric) Outer 281 mm
For Use With (Application) For use with distilled water and culture media.
Graduations Molded-in graduations in 1 gal. and 5 L increments
Height (Metric) With Closure 528 mm
Includes Molded-in handles
Material Polypropylene
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Carboy with Tubulation
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.