missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene, Carboy with 3 in. Sanitary Neck
GREENER_CHOICE

Catalog No. 029638A
Change view
Click to view available options
Capacity (Metric):
10 L
20 L
50 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
029638A 10 L
029638B 20 L
029638C 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 029638A Supplier Thermo Scientific™ Supplier No. 26300010

For Use With: Perform aseptic fluid transfer or sampling in biopharmaceutical applications; Autoclavable; Color: White

Perform aseptic fluid transfer or sampling in biopharmaceutical applications with Thermo Scientific™ Nalgene™ Polypropylene Sanitary Carboys with 3 in. Sanitary Neck Design. The carboy's welded sanitary neck is easy to clean with no screw threads.

  • Versatile with 3 in. sanitary tri-clover flange
  • Sanitary design offers fewer surfaces for contamination
  • Autoclavable
  • Leakproof†

Specifications

Autoclavable Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Closure Size 3 in.
Color White
Diameter (Metric) Outer 250 mm
For Use With (Application) Sampling, Storage, Transportation ​
Height (Metric) With Closure 353 mm
Includes Handles
Material Polypropylene
Neck Style 3 in. Sanitary Neck Design
No. per Case 1 Ea.
Product Type Carboy with Sanitary Neck
Sterility Non-sterile
Workflow Step Upstream media/buffer prep and storage
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.