missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels

Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.

Supplier:  Thermo Scientific™ 42500055

 View more versions of this product

Catalog No. 10-347B



Get 1 Extra Nalgene Product with Purchase
View all promotions
Only null left

Description

Description

Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.

  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable
Specifications

Specifications

54 mm
7 mm
Autoclavable
61 mm
41 mL
Polypropylene
Funnel Analytical PP 55mm
12/Pk.; 36/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions