missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
GREENER_CHOICE

Catalog No. 10347H
Change view
Click to view available options
Diameter (Metric)Top:
35 mm
54 mm
63 mm
76 mm
98 mm
158 mm
Capacity (Metric):
10 mL
41 mL
64 mL
114 mL
254 mL
976 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)Top Capacity (Metric)
10347H 158 mm 976 mL
10347C 35 mm 10 mL
10347B 54 mm 41 mL
10347D 63 mm 64 mL
10347E 76 mm 114 mL
10347G 98 mm 254 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 10347H Supplier Thermo Scientific™ Supplier No. 42500160

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.

Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.

  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable

Specifications

Diameter (Metric)Top 158 mm
Capacity (Metric) 976 mL
Diameter (Metric) Stem 13 mm
Material Polypropylene
Autoclavable Autoclavable
Description Funnel Analytical PP 160mm
Length (Metric) Stem 152 mm
Quantity 2/Pk.; 12/Cs.