missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels

Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.
Supplier: Thermo Scientific™ 42500065
Description
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.
- Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
- Autoclavable
Chemical Identifiers
| Polypropylene |
Specifications
| 63 mm | |
| 7 mm | |
| Autoclavable | |
| 58 mm |
| 64 mL | |
| Polypropylene | |
| Funnel Analytical PP 65mm | |
| 12/Pk.; 36/Cs. |