missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
GREENER_CHOICE

Catalog No. 10347C
Change view
Click to view available options
Diameter (Metric)Top:
35 mm
54 mm
63 mm
76 mm
98 mm
158 mm
Capacity (Metric):
10 mL
41 mL
64 mL
114 mL
254 mL
976 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)Top Capacity (Metric)
10347C 35 mm 10 mL
10347B 54 mm 41 mL
10347D 63 mm 64 mL
10347E 76 mm 114 mL
10347G 98 mm 254 mL
10347H 158 mm 976 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 10347C Supplier Thermo Scientific™ Supplier No. 42500035

Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.

Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.

  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable

Specifications

Diameter (Metric)Top 35 mm
Capacity (Metric) 10 mL
Diameter (Metric) Stem 6 mm
Material Polypropylene
Autoclavable Autoclavable
Description Funnel Analytical PP 35mm
Length (Metric) Stem 50 mm
Quantity 12/Pk.; 36/Cs.