missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polyolefin Right-To-Know Custom Labeling System
GREENER_CHOICE

Catalog No. 1338260
Change view
Click to view available options
Height (Metric):
76.2 mm
Width (Metric):
127 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Width (Metric)
1338260 76.2 mm 127 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1338260 Supplier Thermo Scientific™ Supplier No. 63161000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Create labels that identify a chemical and its properties using the Thermo Scientific™ Nalgene™ Polyolefin Right-To-Know Custom Labeling System.

Create labels that identify a chemical and its properties using the Thermo Scientific™ Nalgene™ Polyolefin Right-To-Know Custom Labeling System.

Features:
  • Overlay provided to protect label from chemical contact
  • Complete instructions included with each package
  • Hazard symbol stickers are GHS-compliant
Note: Refer to the appropriate Material Safety Data Sheet (MSDS) or other reference material to verify the correct codes required for accurate and safe use.

Specifications

Description Custom Right-To-Know Label System
Color Assorted
Format Label
Material Polyolefin
No. per Case 150
No. per Pack 25
Width (English) 5 in.
Width (Metric) 127 mm
Height (English) 3 in.
Height (Metric) 76.2 mm