missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels

Combine these waterproof, chemical resistant labels with photocopiers, laser printers or inkjet printers. Will not wrinkle, curl or melt.

$1719.47 - $2060.19

Specifications

Material Polyolefin
Product Type Label
View More Specs

Products 5
Catalog Number Mfr. No. Height (Metric) Width (Metric) Format Price Qty  
Catalog Number Mfr. No. Height (Metric) Width (Metric) Format Price Qty  
02-923-280
View Documents
Thermo Scientific™
63190010
22 mm 22 mm Case
Case of 4000 Each for $2,009.59
Only null left
 
02-923-281
View Documents
Thermo Scientific™
63190015
20 mm 38 mm Case
Case of 3250 Each for $1,860.06
Only null left
 
02-923-282
View Documents
Thermo Scientific™
63190020
25 mm 67 mm Case
Case of 1500 Each for $1,788.69
Only null left
 
02-923-283
View Documents
Thermo Scientific™
63190030
13 mm 44 mm Case
Case of 4000 Each for $2,060.19
Only null left
 
02-923-284
View Documents
Thermo Scientific™
63190040
51 mm 102 mm Case
Case of 500 Each for $1,719.47
Only null left
 
Description

Description

Combine Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels with photocopiers, laser printers or inkjet printers. Waterproof and compatible with most lab chemicals, Nalgene polyolefin labels will not wrinkle, curl or melt. They accept a variety of inks including copier, marker, ballpoint, gels and technical markers.

Durable Polyolefin Labels

  • Rated to −29°C
  • Resistant to ultraviolet light, mildew, chemicals and dirt and won't discolor, crack or tear
  • Excellent for labeling microscopic slides— adhesive backing sticks well to any plastic, glass or metal surface
  • Chemically compatible with most lab reagents although solvents may dissolve adhesive
  • Autoclavable
  • Formatted for easy setup in word processing programs; instructions included in each pack

Note: For use with all writing devices except pencil. Test specific inks for suitability prior to use.

.

Specifications

Specifications

Polyolefin
Label
Videos
SDS
Documents

Documents