missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate, Carboy with Sanitary Neck
GREENER_CHOICE

Catalog No. 0296120
Change view
Click to view available options
Quantity:
Case of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0296120 Case of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0296120 Supplier Thermo Scientific™ Supplier No. 22610050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use With: Designed for use as receiver or dispensing vessel in biopharmaceutical applications; Autoclavable; Color: Clear

Clean the one-piece molded, non-threaded Thermo Scientific™ Nalgene™ Polycarbonate Sanitary Carboy more easily than threaded containers. Designed for use as receiver or dispensing vessel in biopharmaceutical applications, the 3 in. sanitary flange molded on the neck accepts standard flange fittings. A clamping closure system securely seals and will not back off.

  • Solid ported sanitary end caps are available
  • Made of same polycarbonate (PC) resin as Nalgene PC Clearboys and PC Bottles, permitting switch to Sanitary PC Carboys without material validation issues
  • To seal closure, use with Nalgene Heavy Duty Clamp (Cat. No. 2865-0300) and Gasket (Cat. No. 2672-0300)
  • Autoclavable

Specifications

Autoclavable Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Size 3 in.
Color Clear
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
Includes Sanitary Neck
Material Polycarbonate
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Carboy with Sanitary Neck
Sterility Non-sterile
Volume (English) Brim 6.3 gal.
Volume (Metric) Brim 24 L
Quantity Case of 4
Workflow Step Upstream media/buffer prep and storage. Upstream process solution storage
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.