Learn More
Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure
Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.
Supplier: Thermo Scientific™ 41090250
Description
Perform titrations, preparation and storage of culture media with Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure, which provide a combination of chemical resistance, strength and transparency. Rugged, shatter-proof construction combined with light weight make these flasks a valuble element of lab safety programs.
Features:- Excellent chemical resistance to a wide variety of lab reagents
- More transparent than polypropylene but with similar chemical resistance
- Ideal for preparation and storage of culture media and titrations
- All sizes have easy-to-read graduations
- Autoclavable/Graduated
Includes:Polypropylene screw closures
Specifications
| 8 oz. | |
| 250 mL | |
| 12 | |
| Autoclavable | |
| Yes | |
| Non-sterile | |
| Screw |
| Flask Erlenmeyer Screw Closure PMP 250mL | |
| PMP | |
| 4 | |
| Reusable | |
| Narrow | |
| Plain | |
| 38 mm |