Learn More
Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure
Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.
Supplier: Thermo Scientific™ 41090125
Description
Perform titrations, preparation and storage of culture media with Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure, which provide a combination of chemical resistance, strength and transparency. Rugged, shatter-proof construction combined with light weight make these flasks a valuble element of lab safety programs.
Features:- Excellent chemical resistance to a wide variety of lab reagents
- More transparent than polypropylene but with similar chemical resistance
- Ideal for preparation and storage of culture media and titrations
- All sizes have easy-to-read graduations
- Autoclavable/Graduated
Includes:Polypropylene screw closures
Specifications
| 4 oz. | |
| 125 mL | |
| 24 | |
| Autoclavable | |
| Yes | |
| Non-sterile | |
| Screw |
| Flask Erlenmeyer Screw Closure PMP 125mL | |
| PMP | |
| 6 | |
| Reusable | |
| Narrow | |
| Plain | |
| 33 mm |