missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure
GREENER_CHOICE

Catalog No. 100425J
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Closure Size:
33 mm
38 mm
43 mm
53 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
100425J 1000 mL 53 mm
100425C 125 mL 33 mm
100425E 250 mL 38 mm
100425G 500 mL 43 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 100425J Supplier Thermo Scientific™ Supplier No. 41091000
Get a Water Bottle with Plastics Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.

Perform titrations, preparation and storage of culture media with Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure, which provide a combination of chemical resistance, strength and transparency. Rugged, shatter-proof construction combined with light weight make these flasks a valuble element of lab safety programs.

Features:
  • Excellent chemical resistance to a wide variety of lab reagents
  • More transparent than polypropylene but with similar chemical resistance
  • Ideal for preparation and storage of culture media and titrations
  • All sizes have easy-to-read graduations
  • Autoclavable/Graduated

Includes:Polypropylene screw closures

Specifications

Capacity (English) 32 oz.
Description Flask Erlenmeyer Screw Closure PMP 1000mL
Capacity (Metric) 1000 mL
Material PMP
No. per Case 6
No. per Pack 2
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Plain
Closure Type Screw
Closure Size 53 mm
Show More Show Less