missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PFA Graduated Plastic Beakers with Handles
GREENER_CHOICE

Conduct high-purity applications that use strong oxidizing agents, acids, hydrocarbons and ketones with these PFA graduated beakers with handles.

Supplier:  Thermo Scientific™ DS15113000

 View more versions of this product

Catalog No. 025925B


Only null left

Alert:

Not for use on hotplates.


Description

Description

Conduct high-purity applications that use strong oxidizing agents, acids, hydrocarbons and ketones with Thermo Scientific™ Nalgene™ PFA Graduated Beakers with Handles. These beakers feature extraordinary chemical resistance. PFA is non-stick, making the beaker easy to clean and resistant to contamination.

  • Withstands exposure to extreme temperatures from -270° to +250°C
  • Rigid and translucent, enabling easy and consistent viewing of liquid levels
  • Molded-in graduation in both oz. and mL won't wear off with continuous use or washings
  • Excellent chemical resistance to strong ozidizing agents, acids, hydrocarbons and ketones make these beakers useful for a broad range of high-purity applications
  • Molded-in rectangular handle for easy, safe handling and pouring
  • Autoclavable
Specifications

Chemical Identifiers

Natural translucent
Beaker
Reusable
Molded Dual Scale
PFA
Autoclavable
Yes
Handled

Specifications

Natural translucent
3000 mL
Beaker
Reusable
Molded Dual Scale
Beaker with Handle PFA 3000 mL
PFA
Autoclavable
Yes
Handled
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions