missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination by using PETG serum vials with crimp finish. Lighter in weight than glass, these vials are made for use with stoppers and caps.
$5,575.40
Specifications
| Description | Serum Vials |
|---|---|
| Color | Clear |
| Material | PETG |
| Sterility | Sterile |
| Product Line | Nalgene |
| Catalog Number | Mfr. No. | Volume (Metric) | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Closure Size | Price | Quantity & Availability | ||||
14178177
![]() |
Thermo Scientific™
3420300003 |
3 mL | 13 mm |
Case of 3451 Each for $5,575.40
|
|
||||
Description
Protect against leakage, breakage, and contamination. Thermo Scientific™ Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.
- PETG
- Non-cytotoxic
- Non-pyrogenic
- Packaged in trayless shrink-wrap modules
- USP VI-compliant
- Sterile to 10-6 SAL
Ordering Information:
Serum Vials are packed in trayless shrink-wrapped modules. Stoppers and Caps not included.
Specifications
| Serum Vials | |
| PETG | |
| Nalgene | |
| Stopper, Cap | |
| Serum Vial |
| Clear | |
| Sterile | |
| Stoppers and Caps (Not Included) | |
| ISO 13485:2016 |