missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
GREENER_CHOICE

Catalog No. 03313801
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
03313801 10 mL
03305082 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03313801 Supplier Thermo Scientific™ Supplier No. 3420320010
  • ONLINE EXCLUSIVE PRICE
    $2,198.20
    $1,022.78 / Case of 1260

    Save $1,175.42

    53.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protect against leakage, breakage and contamination with these PETG serum vials. Lighter in weight than glass, they are made for use with lined continuous-thread closures (available separately).

Protect against leakage, breakage, and contamination with Thermo Scientific™ Nalgene PETG Serum Vials. These serum vials are lighter in weight than glass, and are made for use with lined continuous-thread closures (available separately).

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL
  • Leakproof† with Nalgene lined continuous-thread closures (Cat. No. 342158)

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 342158).

Specifications

Description Serum Vials
Diameter (Metric) Inner 11 mm
Color Clear
Diameter (Metric) Outer 24 mm
Material PETG
Sterility Sterile
Volume (Metric) 10 mL
No. per Case 1260
No. per Pack 252
Volume (English) 1/3 oz.
Product Line Nalgene
Volume (Metric) Total 10 mL
Closure Type Lined Continuous-thread Closure (Sold Separately)
Quantity 1260 Cases, 252 Pk.
Closure Size 20-415
For Use With (Equipment) Lined Continuous-Thread Closure
Certifications/Compliance ISO 13485:2016
Height (Metric) 54 mm
Product Type Serum Vial
Show More Show Less