missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
GREENER_CHOICE

Catalog No. 03305082
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
03305082 5 mL
03313801 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03305082 Supplier Thermo Scientific™ Supplier No. 3420320005

Protect against leakage, breakage and contamination with these PETG serum vials. Lighter in weight than glass, they are made for use with lined continuous-thread closures (available separately).

Protect against leakage, breakage, and contamination with Thermo Scientific™ Nalgene PETG Serum Vials. These serum vials are lighter in weight than glass, and are made for use with lined continuous-thread closures (available separately).

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL
  • Leakproof† with Nalgene lined continuous-thread closures (Cat. No. 342158)

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 342158).

Specifications

Description Serum Vials
Diameter (Metric) Inner 11 mm
Color Clear
Diameter (Metric) Outer 22 mm
Material PETG
Sterility Sterile
Volume (Metric) 5 mL
No. per Case 1656
No. per Pack 276
Volume (English) 1/6 oz.
Product Line Nalgene
Volume (Metric) Total 5 mL
Closure Type Lined Continuous-thread Closure (Sold Separately)
Quantity 1656 Cases, 276 Pk.
Closure Size 20-415
For Use With (Equipment) Lined Continuous-Thread Closure
Certifications/Compliance ISO 13485:2016
Height (Metric) 44 mm
Product Type Serum Vial
Show More Show Less