missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
GREENER_CHOICE

Catalog No. 14178176
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14178176 20 mL
03316 5 mL
03317 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14178176 Supplier Thermo Scientific™ Supplier No. 3220320020
  • ONLINE EXCLUSIVE PRICE
    $1,381.00
    $560.69 / Case of 612 Each

    Save $820.31

    59.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protect against leakage, breakage and contamination by using PETG nonsterile serum vials with continuous thread closures.

Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Nonsterile
  • Leakproof† with Nalgene lined continuous-thread closures (available separately)

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).

Specifications

Description Serum Vials
Diameter (Metric) Inner 11 mm
Color Clear
Diameter (Metric) Outer 30 mm
Material PETG
Sterility Non-sterile
Volume (Metric) 20 mL
No. per Case 612
No. per Pack 153
Volume (English) 2/3 oz.
Product Line Nalgene
Volume (Metric) Total 20 mL
Closure Type Continuous-thread Closure (Sold Separately)
Quantity 153 Pk., 612 Cases
Closure Size 20-415
For Use With (Equipment) Lined Continuous Thread Closure
Certifications/Compliance ISO 13485:2003
Height (Metric) 64 mm
Product Type Serum Vial
Show More Show Less