missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
GREENER_CHOICE

Catalog No. 03316
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
03316 5 mL
03317 10 mL
14178176 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03316 Supplier Thermo Scientific™ Supplier No. 3220320005

Protect against leakage, breakage and contamination by using PETG nonsterile serum vials with continuous thread closures.

Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Nonsterile
  • Leakproof† with Nalgene lined continuous-thread closures (available separately)

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).

Specifications

Description Serum Vials
Diameter (Metric) Inner 11 mm
Color Clear
Diameter (Metric) Outer 22 mm
Material PETG
Sterility Non-sterile
Volume (Metric) 5 mL
No. per Case 1656
No. per Pack 276
Volume (English) 1/6 oz.
Product Line Nalgene
Volume (Metric) Total 5 mL
Closure Type Screw Cap (Not Included)
Quantity 276 Pk. , 1656 Cases
Closure Size 20-415
For Use With (Equipment) Lined Continuous Thread Closure
Certifications/Compliance ISO 13485:2003
Height (Metric) 44 mm
Product Type Serum Vial
Show More Show Less